Learn More
Electron Microscopy Sciences FlowView Starter Kit - Semi
SDP
Catalog No. 501928103
Encompass
FlowView starter kit provides world-leading innovative liquid sample inspection with SEM. These economic and easy-to-use starter kits include stage, accessories, and 24 Microscopic Fluid Chips. The chips are compatible with SEM for most major brands. Kit contains tweezer, pipette, torque wrench, carrier box, storage box, and 24 microscopic fluid chips. Use for CMP slurry particles, wet etching, photoresist developing, electroplating. Tweezer, pipette, torque wrench, carrier box, storage box, 24 microscopic fluid chips.
Catalog No. 50-192-8103 Supplier Electron Microscopy Sciences Supplier No. FV103
FlowView starter kit provides world-leading innovative liquid sample inspection with SEM. These economic and easy-to-use starter kits include stage, accessories, and 24 Microscopic Fluid Chips. The chips are compatible with SEM for most major brands. Kit contains tweezer, pipette, torque wrench, carrier box, storage box, and 24 microscopic fluid chips. Use for CMP slurry particles, wet etching, photoresist developing, electroplating. Tweezer, pipette, torque wrench, carrier box, storage box, 24 microscopic fluid chips.