Learn More
Electron Microscopy Sciences FlowView Starter Kit - Standard
SDP
Supplier:  Electron Microscopy Sciences FV102
Encompass
FlowView starter kit provides world-leading innovative liquid sample inspection with SEM. These economic and easy-to-use starter kits include stage, accessories, and 24 Microscopic Fluid Chips. The chips are compatible with SEM for most major brands. Kit contains tweezer, pipette, torque wrench, carrier box, storage box, and 24 microscopic fluid chips. For most sample droplets (paint, mixing slurry, particles in oil). Tweezer, pipette, torque wrench, carrier box, storage box, 24 microscopic fluid chips.
Catalog No. 50-192-8102
Add to Cart
Request a Quote