Learn More
Medchemexpress LLC HY-B0097 100mg Medchemexpress, Floxuridine CAS:50-91-9 Purity:>98%
SDP
Catalog No. 502031025
Encompass_Preferred
Medchemexpress, HY-B0097 100mg Floxuridine CAS:50-91-9 Floxuridine (5-Fluorouracil 2'-deoxyriboside) is a pyrimidine analog and known as an oncology antimetabolite. Floxuridine inhibits Poly(ADP-Ribose) polymerase and induces DNA damage by activating the ATM and ATR checkpoint signaling pathways in vitro. Floxuridine is a extreamly potent inhibitor for S. aureus infection and induces cell apoptosis[2]. Floxuridine has antiviral effects against HSV and CMV[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1025 Supplier Medchemexpress LLC Supplier No. HYB0097100MG
Medchemexpress, HY-B0097 100mg Floxuridine CAS:50-91-9 Floxuridine (5-Fluorouracil 2'-deoxyriboside) is a pyrimidine analog and known as an oncology antimetabolite. Floxuridine inhibits Poly(ADP-Ribose) polymerase and induces DNA damage by activating the ATM and ATR checkpoint signaling pathways in vitro. Floxuridine is a extreamly potent inhibitor for S. aureus infection and induces cell apoptosis[2]. Floxuridine has antiviral effects against HSV and CMV[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.