Learn More
Apexbio Technology LLC Floxuridine, 25mg. Cas: 50-91-9 MFCD: MFCD00006530. Antineoplastic antimetabolite
SDP
Supplier:  Apexbio Technology LLC A240225
Encompass_Preferred
Antineoplastic antimetabolite. Exhibits antiproliferative activity; inhibits thymidylate synthetase and disrupts DNA replication in human cells. Induces double-strand DNA breaks; activates ATR and ATM signaling pathways. Induces phosphorylation of Chk1 an Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4741