Learn More
Medchemexpress LLC Floxuridine | 50-91-9 | 99.97% | C9H11FN2O5 | 200 MG
SDP
Catalog No. 5000432880
Encompass_Preferred
Floxuridine is a pyrimidine analog and an oncology antimetabolite. It inhibits Poly(ADP-Ribose) polymerase and induces DNA damage by activating ATM and ATR checkpoint signaling pathways. Floxuridine is a potent inhibitor for S. aureus infection and induces cell apoptosis, in addition to having antiviral effects against HSV and CMV.
  • Inhibits Poly(ADP-Ribose) polymerase
  • Induces DNA damage by activating ATM and ATR checkpoint signaling pathways
  • Acts as a potent inhibitor for S. aureus infection
  • Induces cell apoptosis
  • Shows antiviral effects against HSV and CMV
  • Potentiates the sensitivity of ovarian cancer cells when co-exposed with PARP inhibitors
  • Causes G1/S-phase arrest in ovarian cancer cells
  • Reduces the virulence of Staphylococcus aureus
Catalog No. 50-004-32880 Supplier Medchemexpress LLC Supplier No. HYB0097200MG
Floxuridine is a pyrimidine analog and an oncology antimetabolite. It inhibits Poly(ADP-Ribose) polymerase and induces DNA damage by activating ATM and ATR checkpoint signaling pathways. Floxuridine is a potent inhibitor for S. aureus infection and induces cell apoptosis, in addition to having antiviral effects against HSV and CMV.
  • Inhibits Poly(ADP-Ribose) polymerase
  • Induces DNA damage by activating ATM and ATR checkpoint signaling pathways
  • Acts as a potent inhibitor for S. aureus infection
  • Induces cell apoptosis
  • Shows antiviral effects against HSV and CMV
  • Potentiates the sensitivity of ovarian cancer cells when co-exposed with PARP inhibitors
  • Causes G1/S-phase arrest in ovarian cancer cells
  • Reduces the virulence of Staphylococcus aureus