Learn More
Lkt Laboratories Floxuridine, ≥98%, 1GR
SDP
Supplier:  Lkt Laboratories F45571G
Encompass_Preferred
5-Fluorouracil derivative, fluorinated pyrimidine nuceloside analog; thymidylate synthase inhibitor. CAS: 50-91-9. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4920