Learn More
McKesson General Medical Sysmex Fluorocell™ WDF, Hematology Reagent for WBC Counting, 2 x 42 mL
Catalog No. NC2143722
Encompass
The Sysmex Fluorocell™ WDF is a liquid hematology reagent specifically designed for accurate white blood cell counting and differentiation on Sysmex automated analyzers. Supplied in a dual 42 mL cassette format, this specialized staining reagent ensures reliable and precise WBC subpopulation analysis, aiding laboratory professionals in obtaining trusted hematology results.
This item is not returnable. View return policy
Catalog No. NC2143722 Supplier McKesson General Medical Supplier No. 1195265
The Sysmex Fluorocell™ WDF is a liquid hematology reagent specifically designed for accurate white blood cell counting and differentiation on Sysmex automated analyzers. Supplied in a dual 42 mL cassette format, this specialized staining reagent ensures reliable and precise WBC subpopulation analysis, aiding laboratory professionals in obtaining trusted hematology results.