Learn More
Ace Glass, Inc. Flask, round bottom, 100mL, 14/20, 0-4mm Hi-Vac valve sidearm with 8mm septa port, No-Air
SDP
Catalog No. 50979452
Encompass_Preferred
100 mL round bottom borosilicate glass flask with standard taper center neck and a 0-4 Hi-Vac valve for precise control. Flask is IDeal for simple NO-AIR and vacuum applications. Outlet arm is 8MM and can be fitted with a rubber septa for insertion of samples or additives via syringe.
Catalog No. 50-979-452 Supplier Ace Glass, Inc. Supplier No. 776404
100 mL round bottom borosilicate glass flask with standard taper center neck and a 0-4 Hi-Vac valve for precise control. Flask is IDeal for simple NO-AIR and vacuum applications. Outlet arm is 8MM and can be fitted with a rubber septa for insertion of samples or additives via syringe.