Learn More
Chemglass Life Sciences Low Form Dewar Flask, Cylindrical, 150mL Capacity, Fits 100mL Round Bottom Flask (RBF) , 80mm ID X 35mm Inside Depth, 101mm OD X 66mm Height
SDP
Catalog No. CG159201
Encompass_Preferred
Low Form Dewar Flask, Cylindrical, 150mL Capacity, Fits 100mL RBF, 80mm ID X 35mm Inside Depth, 101mm OD X 66mm Height. Similar to CG-1590, but with a semi-flat bottom which increases coolant capacity and permits magnetic stirring of contents of vessel. With aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.
Catalog No. CG159201 Supplier Chemglass Life Sciences Supplier No. CG159201
Low Form Dewar Flask, Cylindrical, 150mL Capacity, Fits 100mL RBF, 80mm ID X 35mm Inside Depth, 101mm OD X 66mm Height. Similar to CG-1590, but with a semi-flat bottom which increases coolant capacity and permits magnetic stirring of contents of vessel. With aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.