Learn More
Chemglass Life Sciences Low Form Dewar Flask, Cylindrical, 350mL Capacity, Fits 100mL Round Bottom Flask (RBF) , 80mm ID X 75mm Inside Depth, 101mm OD X 117mm Height
SDP
Catalog No. CG159202
Encompass_Preferred
Low Form Dewar Flask, Cylindrical, 350mL Capacity, Fits 100mL RBF, 80mm ID X 75mm Inside Depth, 101mm OD X 117mm Height. Similar to CG-1590, but with a semi-flat bottom which increases coolant capacity and permits magnetic stirring of contents of vessel. With aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.
Catalog No. CG159202 Supplier Chemglass Life Sciences Supplier No. CG159202
Low Form Dewar Flask, Cylindrical, 350mL Capacity, Fits 100mL RBF, 80mm ID X 75mm Inside Depth, 101mm OD X 117mm Height. Similar to CG-1590, but with a semi-flat bottom which increases coolant capacity and permits magnetic stirring of contents of vessel. With aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.