Learn More
Chemglass Life Sciences Low Form Dewar Flask, Cylindrical, 850mL Capacity, Fits 500mL Round Bottom Flask (RBF) , 130mm ID X 75mm Inside Depth, 160mm OD X 110mm Height
SDP
Catalog No. CG159203
Encompass_Preferred
Low Form Dewar Flask, Cylindrical, 850mL Capacity, Fits 500mL RBF, 130mm ID X 75mm Inside Depth, 160mm OD X 110mm Height. Similar to CG-1590, but with a semi-flat bottom which increases coolant capacity and permits magnetic stirring of contents of vessel. With aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.
Catalog No. CG159203 Supplier Chemglass Life Sciences Supplier No. CG159203
Low Form Dewar Flask, Cylindrical, 850mL Capacity, Fits 500mL RBF, 130mm ID X 75mm Inside Depth, 160mm OD X 110mm Height. Similar to CG-1590, but with a semi-flat bottom which increases coolant capacity and permits magnetic stirring of contents of vessel. With aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.