Learn More
Chemglass Life Sciences Dewar Flask, Wide Mouth, Metal Housing, 4300mL Capacity, 143mm ID X 305mm Inside Depth, 178mm OD X 365mm Height
SDP
Catalog No. CG159405
Encompass_Preferred
Dewar Flask, Wide Mouth, Metal Housing, 4300mL Capacity, 143mm ID X 305mm Inside Depth, 178mm OD X 365mm Height. Similar to CG-1593, but with a full length aluminum housing which extends to the top rim for added protection. Supplied complete with a vented polyethylene stopper to minimize evaporation.
Catalog No. CG159405 Supplier Chemglass Life Sciences Supplier No. CG159405
Dewar Flask, Wide Mouth, Metal Housing, 4300mL Capacity, 143mm ID X 305mm Inside Depth, 178mm OD X 365mm Height. Similar to CG-1593, but with a full length aluminum housing which extends to the top rim for added protection. Supplied complete with a vented polyethylene stopper to minimize evaporation.