Learn More
Chemglass Life Sciences Low Form Dewar Flask, Hemispherical, 150mL Capacity, Fits 100mL Round Bottom Flask (RBF) , 80mm ID X 40mm Inside Depth, 106mm OD X 66mm Height
SDP
Catalog No. CG159001
Encompass_Preferred
Low Form Dewar Flask, Hemispherical, 150mL Capacity, Fits 100mL RBF, 80mm ID X 40mm Inside Depth, 106mm OD X 66mm Height. Low form hemispherical Dewar with protective aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.
Catalog No. CG159001 Supplier Chemglass Life Sciences Supplier No. CG159001
Low Form Dewar Flask, Hemispherical, 150mL Capacity, Fits 100mL RBF, 80mm ID X 40mm Inside Depth, 106mm OD X 66mm Height. Low form hemispherical Dewar with protective aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.