Learn More
Chemglass Life Sciences Low Form Dewar Flask, Hemispherical, 275mL Capacity, Fits 250mL Round Bottom Flask (RBF) , 100mm ID X 55mm Inside Depth, 132mm OD X 75mm Height
SDP
Catalog No. CG159002
Encompass_Preferred
Low Form Dewar Flask, Hemispherical, 275mL Capacity, Fits 250mL RBF, 100mm ID X 55mm Inside Depth, 132mm OD X 75mm Height. Low form hemispherical Dewar with protective aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.
Catalog No. CG159002 Supplier Chemglass Life Sciences Supplier No. CG159002
Low Form Dewar Flask, Hemispherical, 275mL Capacity, Fits 250mL RBF, 100mm ID X 55mm Inside Depth, 132mm OD X 75mm Height. Low form hemispherical Dewar with protective aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.