Learn More
Wilmad Labglass FLSK DWR CYL LAB GRADE 350ML
Catalog No. 50800774
Encompass
Dewar Flask, Cylindrical, Laboratory Grade, Completely Silvered, w/ Aluminum Base. Capacity: 350mL. Dimensions: Inside Diameter: 2- 3/8 in. Inside Depth: 4- 7/16 in.
Catalog No. 50-800-774 Supplier Wilmad Labglass Supplier No. LG7671100
Dewar Flask, Cylindrical, Laboratory Grade, Completely Silvered, w/ Aluminum Base. Capacity: 350mL. Dimensions: Inside Diameter: 2- 3/8 in. Inside Depth: 4- 7/16 in.