Learn More
Wilmad Labglass FLSK DWR CYL LAB GRADE 4.3L
Catalog No. 50801793
Encompass
Dewar Flask, Cylindrical, Laboratory Grade, Completely Silvered, w/ Aluminum Base. Capacity: 4300mL. Dimensions: Inside Diameter: 6 in. Inside Depth: 10- 7/8 in.
Catalog No. 50-801-793 Supplier Wilmad Labglass Supplier No. LG7671108
Dewar Flask, Cylindrical, Laboratory Grade, Completely Silvered, w/ Aluminum Base. Capacity: 4300mL. Dimensions: Inside Diameter: 6 in. Inside Depth: 10- 7/8 in.