Learn More
Wilmad Labglass Dewar Flask, Cylindrical, 665 mL, Laboratory Grade
Catalog No. 50800454
Encompass
Dewar Flask, Cylindrical, Laboratory Grade, Completely Silvered, w/ Aluminum Base. Capacity: 665mL. Dimensions: Inside Diameter: 2- 3/4 in. Inside Depth: 7- 11/16 in.
Catalog No. 50-800-454 Supplier Wilmad Labglass Supplier No. LG7671102
Dewar Flask, Cylindrical, Laboratory Grade, Completely Silvered, w/ Aluminum Base. Capacity: 665mL. Dimensions: Inside Diameter: 2- 3/4 in. Inside Depth: 7- 11/16 in.