Learn More
Wilmad Labglass FLSK DWR LOW CYL 350ML
Catalog No. 50800995
Encompass
Dewar Flask, Cylindrical, Low Form, Completely Silvered, w/ Base. Capacity: 350mL. Dimensions: Inside Diameter: 80mm; Inside Depth: 75mm.
Catalog No. 50-800-995 Supplier Wilmad Labglass Supplier No. LG7680102
Dewar Flask, Cylindrical, Low Form, Completely Silvered, w/ Base. Capacity: 350mL. Dimensions: Inside Diameter: 80mm; Inside Depth: 75mm.