Learn More
Wilmad Labglass FLSK DWR LOW CYL 850ML
Catalog No. 50801650
Encompass
Dewar Flask, Cylindrical, Low Form, Completely Silvered, w/ Base. Capacity: 850mL. Dimensions: Inside Diameter: 130mm; Inside Depth: 75mm.
Catalog No. 50-801-650 Supplier Wilmad Labglass Supplier No. LG7680106
Dewar Flask, Cylindrical, Low Form, Completely Silvered, w/ Base. Capacity: 850mL. Dimensions: Inside Diameter: 130mm; Inside Depth: 75mm.