Learn More
Chemglass Life Sciences 500mL Filtering Flask, Heavy Wall, 24/40 Outer Joint
SDP
Catalog No. CG154903
Encompass_Preferred
500mL Filtering Flask, Heavy Wall, 24/40 Outer Joint. HEAVY WALL Erlenmeyer shaped filtering flask with standard taper outer joint. For stoppers see CG-3000.
Catalog No. CG154903 Supplier Chemglass Life Sciences Supplier No. CG154903
500mL Filtering Flask, Heavy Wall, 24/40 Outer Joint. HEAVY WALL Erlenmeyer shaped filtering flask with standard taper outer joint. For stoppers see CG-3000.