Learn More
Wilmad Labglass FLSK RB 2N 100ML 24/40C 24/40S
Catalog No. 50806708
Encompass_Preferred
Boiling Flask, Round Bottom, 2-Neck, 100mL capacity. Joint Sizes: Center Neck: [ST] 24/40 outer; Angled Side Neck: [ST] 24/40 outer. Side neck is angled 20degree to the center neck.
This item is not returnable. View return policy
Catalog No. 50-806-708 Supplier Wilmad Labglass Supplier No. LG7291222
Boiling Flask, Round Bottom, 2-Neck, 100mL capacity. Joint Sizes: Center Neck: [ST] 24/40 outer; Angled Side Neck: [ST] 24/40 outer. Side neck is angled 20degree to the center neck.