Learn More
Wilmad Labglass FLSK RB 2N 250ML 24/40C 24/40S
Catalog No. 50803251
Encompass_Preferred
Boiling Flask, Round Bottom, 2-Neck, 250mL capacity. Joint Sizes: Center Neck: [ST] 24/40 outer; Angled Side Neck: [ST] 24/40 outer. Side neck is angled 20degree to the center neck.
Catalog No. 50-803-251 Supplier Wilmad Labglass Supplier No. LG7291234
Boiling Flask, Round Bottom, 2-Neck, 250mL capacity. Joint Sizes: Center Neck: [ST] 24/40 outer; Angled Side Neck: [ST] 24/40 outer. Side neck is angled 20degree to the center neck.