Learn More
Wilmad Labglass FLSK RB 3N 100ML 14/20C 14/20S
Catalog No. 50803287
Encompass_Preferred
Boiling Flask, Round Bottom, 3-Neck. Capacity: 100mL. Joint Sizes: Center Neck: [ST] 14/20; Side Necks: [ST] 14/20. Round bottom 3-neck boiling flask with vertical center neck and two side necks that are 180degree apart.
Catalog No. 50-803-287 Supplier Wilmad Labglass Supplier No. ML1130712
Boiling Flask, Round Bottom, 3-Neck. Capacity: 100mL. Joint Sizes: Center Neck: [ST] 14/20; Side Necks: [ST] 14/20. Round bottom 3-neck boiling flask with vertical center neck and two side necks that are 180degree apart.