Learn More
Wilmad Labglass FLSK RB 3N 1L 24/40C 24/40S
Catalog No. 50806228
Encompass_Preferred
Boiling Flask, Round Bottom, 3-Neck, 1000mL capacity. Joint Sizes: Center Neck: [ST] 24/40; Vertical Side Necks: [ST] 24/40. Round bottom three neck boiling flask with standard taper joints on the center neck and the vertical side necks.
Catalog No. 50-806-228 Supplier Wilmad Labglass Supplier No. LG7330194
Boiling Flask, Round Bottom, 3-Neck, 1000mL capacity. Joint Sizes: Center Neck: [ST] 24/40; Vertical Side Necks: [ST] 24/40. Round bottom three neck boiling flask with standard taper joints on the center neck and the vertical side necks.