Learn More
Wilmad Labglass FLSK RB 3N 250ML 24/40C 24/40S
Catalog No. 50807048
Encompass_Preferred
Boiling Flask, Round Bottom, 3-Neck, 250mL capacity. Joint Sizes: Center Neck: [ST] 24/40; Angled Side Necks: [ST] 24/40. Round bottom three neck boiling flask with joints on all three necks. Side necks are 180degree apart.
Catalog No. 50-807-048 Supplier Wilmad Labglass Supplier No. LG7331182
Boiling Flask, Round Bottom, 3-Neck, 250mL capacity. Joint Sizes: Center Neck: [ST] 24/40; Angled Side Necks: [ST] 24/40. Round bottom three neck boiling flask with joints on all three necks. Side necks are 180degree apart.