Learn More
Wilmad Labglass FLSK RB 3N 250ML 24/40C 24/40S
Supplier:  Wilmad Labglass LG7330182
Encompass_Preferred
Boiling Flask, Round Bottom, 3-Neck, 250mL capacity. Joint Sizes: Center Neck: [ST] 24/40; Vertical Side Necks: [ST] 24/40. Round bottom three neck boiling flask with standard taper joints on the center neck and the vertical side necks.
Catalog No. 50-807-041