Learn More
Wilmad Labglass FLSK RB 3N 50L 55/50C 45/50S
Catalog No. 50805495
Encompass
Boiling Flask, Round Bottom, 3-Neck, 50 Liter capacity. Joint Sizes: Center Neck: [ST] 55/50; Vertical Side Necks: [ST] 45/50. Round bottom three neck boiling flask with standard taper joints on the center neck and the vertical side necks.
Catalog No. 50-805-495 Supplier Wilmad Labglass Supplier No. LG7330384
Boiling Flask, Round Bottom, 3-Neck, 50 Liter capacity. Joint Sizes: Center Neck: [ST] 55/50; Vertical Side Necks: [ST] 45/50. Round bottom three neck boiling flask with standard taper joints on the center neck and the vertical side necks.