Learn More
Wilmad Labglass FLSK RB 3N 5L 45/50C 24/40S
Catalog No. 50805596
Encompass_Preferred
Boiling Flask, Round Bottom, 3-Neck, 5000mL capacity. Joint Sizes: Center Neck: [ST] 45/50; Vertical Side Necks: [ST] 24/40. Round bottom three neck boiling flask with standard taper joints on the center neck and the vertical side necks.
Catalog No. 50-805-596 Supplier Wilmad Labglass Supplier No. LG7330248
Boiling Flask, Round Bottom, 3-Neck, 5000mL capacity. Joint Sizes: Center Neck: [ST] 45/50; Vertical Side Necks: [ST] 24/40. Round bottom three neck boiling flask with standard taper joints on the center neck and the vertical side necks.