Learn More
Wilmad Labglass FLSK RB 4N 1L 34/45C 24/40S
Catalog No. 50806532
Encompass_Preferred
Boiling Flask, Round Bottom, 4-Neck, 1000mL capacity. Joint Sizes: Center Neck:[ST] 34/45; Vertical Side Necks: [ST] 24/40. Round bottom four neck boiling flask with center neck and three vertical side necks that are 90degree apart.
Catalog No. 50-806-532 Supplier Wilmad Labglass Supplier No. LG7370178
Boiling Flask, Round Bottom, 4-Neck, 1000mL capacity. Joint Sizes: Center Neck:[ST] 34/45; Vertical Side Necks: [ST] 24/40. Round bottom four neck boiling flask with center neck and three vertical side necks that are 90degree apart.