Learn More
Wilmad Labglass FLSK RB 4N 2L 45/50C 24/40S
Catalog No. 50804897
Encompass_Preferred
Boiling Flask, Round Bottom, 4-Neck, 2000mL capacity. Joint Sizes: Center Neck:[ST] 45/50; Vertical Side Necks: [ST] 24/40. Round bottom four neck boiling flask with center neck and three vertical side necks that are 90degree apart.
Catalog No. 50-804-897 Supplier Wilmad Labglass Supplier No. LG7370188
Boiling Flask, Round Bottom, 4-Neck, 2000mL capacity. Joint Sizes: Center Neck:[ST] 45/50; Vertical Side Necks: [ST] 24/40. Round bottom four neck boiling flask with center neck and three vertical side necks that are 90degree apart.