Learn More
Wilmad Labglass FLSK RB 4N 3L 34/45C 24/40S
Catalog No. 50805360
Encompass_Preferred
Boiling Flask, Round Bottom, 4-Neck, 3000mL capacity. Joint Sizes: Center Neck:[ST] 34/45; Vertical Side Necks: [ST] 24/40. Round bottom four neck boiling flask with center neck and three vertical side necks that are 90degree apart.
Catalog No. 50-805-360 Supplier Wilmad Labglass Supplier No. LG7370194
Boiling Flask, Round Bottom, 4-Neck, 3000mL capacity. Joint Sizes: Center Neck:[ST] 34/45; Vertical Side Necks: [ST] 24/40. Round bottom four neck boiling flask with center neck and three vertical side necks that are 90degree apart.