Learn More
Chemglass Life Sciences 250mL Flask, Reaction, 24/40 Outer Joint, 2mm Glass Stpk, Airfree, Schlenk
SDP
Catalog No. AF052804
Encompass_Preferred
250mL Flask, Reaction, 24/40 Outer Joint, 2mm Glass Stpk, Airfree, Schlenk. Round bottom single neck flask having a standard taper outer joint. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.
Catalog No. AF052804 Supplier Chemglass Life Sciences Supplier No. AF052804
250mL Flask, Reaction, 24/40 Outer Joint, 2mm Glass Stpk, Airfree, Schlenk. Round bottom single neck flask having a standard taper outer joint. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.