Learn More
Wilmad Labglass FLSK VOL CL A 1000ML ST22 STPR
Catalog No. 50804936
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 1000mL +/- 0.30mL capacity. Stopper Size: 22.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A(Unserialized flasks) and ASTM E-288.
Catalog No. 50-804-936 Supplier Wilmad Labglass Supplier No. LG8110120
Volumetric Flask, Class A, To Contain, 1000mL +/- 0.30mL capacity. Stopper Size: 22.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A(Unserialized flasks) and ASTM E-288.