Learn More
Wilmad Labglass FLSK VOL CL A 100ML ST13 STPR
Catalog No. 50807752
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 100mL +/- 0.08mL capacity. Stopper Size: 13.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A (Unserialized flasks) and ASTM E-288.
This item is not returnable. View return policy
Catalog No. 50-807-752 Supplier Wilmad Labglass Supplier No. LG8110112
Volumetric Flask, Class A, To Contain, 100mL +/- 0.08mL capacity. Stopper Size: 13.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A (Unserialized flasks) and ASTM E-288.