Learn More
Wilmad Labglass FLSK VOL CL A 10L PLS STPR
Catalog No. 50804601
Encompass
Volumetric Flask, Class A, To Contain, 10000mL +/- 2.5mL capacity.Stopper Size: 38.Calibrated to contain at 20degreeC.Flask is provided with plastic closed-bottom Stopper..
Catalog No. 50-804-601 Supplier Wilmad Labglass Supplier No. LG8116132
Volumetric Flask, Class A, To Contain, 10000mL +/- 2.5mL capacity.Stopper Size: 38.Calibrated to contain at 20degreeC.Flask is provided with plastic closed-bottom Stopper..