Learn More
Wilmad Labglass FLSK VOL CL A 2000ML ST27 STPR
Catalog No. 50803622
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 2000mL +/- 0.50mL capacity. Stopper Size: 27.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A(Unserialized flasks) and ASTM E-288.
Catalog No. 50-803-622 Supplier Wilmad Labglass Supplier No. LG8110122
Volumetric Flask, Class A, To Contain, 2000mL +/- 0.50mL capacity. Stopper Size: 27.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A(Unserialized flasks) and ASTM E-288.