Learn More
Wilmad Labglass FLSK VOL CL A 20L ST38 STPR
Catalog No. 50805221
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 20000mL +/- 4mL capacity.Stopper Size: 38.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A (Unserialized flasks) and ASTM E-288.
Catalog No. 50-805-221 Supplier Wilmad Labglass Supplier No. LG8110134
Volumetric Flask, Class A, To Contain, 20000mL +/- 4mL capacity.Stopper Size: 38.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A (Unserialized flasks) and ASTM E-288.