Learn More
Wilmad Labglass FLSK VOL CL A 250ML ST16 STPR
Catalog No. 50806163
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 250mL +/- 0.12mL capacity. Stopper Size: 16.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A (Unserialized flasks) and ASTM E-288.
Catalog No. 50-806-163 Supplier Wilmad Labglass Supplier No. LG8110116
Volumetric Flask, Class A, To Contain, 250mL +/- 0.12mL capacity. Stopper Size: 16.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A (Unserialized flasks) and ASTM E-288.