Learn More
Wilmad Labglass FLSK VOL CL A 4000ML PLS STPR
Catalog No. 50804030
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 4000mL +/- 1.0mL capacity. Stopper Size: 38.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A(Unserialized flasks) and ASTM E-288.
Catalog No. 50-804-030 Supplier Wilmad Labglass Supplier No. LG8116126
Volumetric Flask, Class A, To Contain, 4000mL +/- 1.0mL capacity. Stopper Size: 38.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A(Unserialized flasks) and ASTM E-288.