Learn More
Wilmad Labglass FLSK VOL CL A 5000ML PLS STPR
Catalog No. 50805822
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 5000mL +/- 1.5mL capacity. Stopper Size: 38.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A(Unserialized flasks) and ASTM E-288.
Catalog No. 50-805-822 Supplier Wilmad Labglass Supplier No. LG8116128
Volumetric Flask, Class A, To Contain, 5000mL +/- 1.5mL capacity. Stopper Size: 38.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A(Unserialized flasks) and ASTM E-288.