Learn More
Wilmad Labglass FLSK VOL CL A 5000ML ST38 STPR
Catalog No. 50806012
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 5000mL +/- 1.5mL capacity. Stopper Size: 38.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A(Unserialized flasks) and ASTM E-288.
Catalog No. 50-806-012 Supplier Wilmad Labglass Supplier No. LG8110128
Volumetric Flask, Class A, To Contain, 5000mL +/- 1.5mL capacity. Stopper Size: 38.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A(Unserialized flasks) and ASTM E-288.