Learn More
Wilmad Labglass FLSK VOL CL A 6000ML ST38 STPR
Catalog No. 50804315
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 6000mL +/- 2.0mL capacity. Stopper Size: 38.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A (Unserialized flasks) and ASTM E-288.
Catalog No. 50-804-315 Supplier Wilmad Labglass Supplier No. LG8110130
Volumetric Flask, Class A, To Contain, 6000mL +/- 2.0mL capacity. Stopper Size: 38.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A (Unserialized flasks) and ASTM E-288.