Learn More
Wilmad Labglass FLSK VOL CL A SRLZD 4ML
Catalog No. 50804208
Encompass
Volumetric Flask, Class A, To Contain, 4mL +/- 0.02mL capacity. Stopper Size: 8. Calibrated to contain at 20degreeC. Flask has serial number and is supplied with a Certificate of Compliance.
Catalog No. 50-804-208 Supplier Wilmad Labglass Supplier No. LG8114106
Volumetric Flask, Class A, To Contain, 4mL +/- 0.02mL capacity. Stopper Size: 8. Calibrated to contain at 20degreeC. Flask has serial number and is supplied with a Certificate of Compliance.