Learn More
Wilmad Labglass FLSK VOL CL A SRLZD 500ML
Catalog No. 50803446
Encompass
Volumetric Flask, Class A, To Contain, 500mL +/- 0.20mL capacity. Stopper Size: 19. Calibrated to contain at 20degreeC. Flask has serial number and is supplied with a Certificate of Compliance.
Catalog No. 50-803-446 Supplier Wilmad Labglass Supplier No. LG8112118
Volumetric Flask, Class A, To Contain, 500mL +/- 0.20mL capacity. Stopper Size: 19. Calibrated to contain at 20degreeC. Flask has serial number and is supplied with a Certificate of Compliance.