Learn More
Wilmad Labglass FLSK VOL KOHLRH CL A 100ML
Catalog No. 50806934
Encompass_Preferred
Volumetric Flask, Kohlrausch, Class A, To Contain, 100ml +/- 0.08mL capacity. Calibrated to contain at 20degreeC. The upper neck of the flask is enlarged to permit easy introduction of solids. Flask has serial number.
Catalog No. 50-806-934 Supplier Wilmad Labglass Supplier No. LG8192100
Volumetric Flask, Kohlrausch, Class A, To Contain, 100ml +/- 0.08mL capacity. Calibrated to contain at 20degreeC. The upper neck of the flask is enlarged to permit easy introduction of solids. Flask has serial number.