Learn More
Wilmad Labglass FLSK VOL KOHLRH CL A 200ML
Catalog No. 50803553
Encompass_Preferred
Volumetric Flask, Kohlrausch, Class A, To Contain, 200ml +/- 0.10mL capacity. Calibrated to contain at 20degreeC. The upper neck of the flask is enlarged to permit easy introduction of solids. Flask has serial number.
Catalog No. 50-803-553 Supplier Wilmad Labglass Supplier No. LG8192102
Volumetric Flask, Kohlrausch, Class A, To Contain, 200ml +/- 0.10mL capacity. Calibrated to contain at 20degreeC. The upper neck of the flask is enlarged to permit easy introduction of solids. Flask has serial number.