Learn More
Eurofins Discoverx Flt-3, active, 10 µg. (14-500)
Supplier:  Eurofins Discoverx 14500
Encompass
N-terminal GST-tagged recombinant, human Flt3 amino acids 564 - end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8873