Learn More
ACROBiosystems ACROBiosystems Human Flt-3 / Flk-2 Protein, Fc Tag
Catalog No. 501956858
Encompass_Preferred
Recombinant Protein;100UG;Human Flt-3, Fc Tag (FL3-H5258) is expressed from human 293 cells (HEK293). It contains AA Asn 27 - Ser 543 (Accession # P36888-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 84.8 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Flt-3 Ligand, Tag Free (Cat. No. FLL-H5218) at 1 μg/mL (100 μL/well) can bind Human Flt-3, Fc Tag (Cat. No. FL3-H5258) with a linear range of 2-31 ng/mL (QC tested).
Catalog No. 50-195-6858 Supplier ACROBiosystems Supplier No. FL3H5258100UG
Recombinant Protein;100UG;Human Flt-3, Fc Tag (FL3-H5258) is expressed from human 293 cells (HEK293). It contains AA Asn 27 - Ser 543 (Accession # P36888-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 84.8 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Flt-3 Ligand, Tag Free (Cat. No. FLL-H5218) at 1 μg/mL (100 μL/well) can bind Human Flt-3, Fc Tag (Cat. No. FL3-H5258) with a linear range of 2-31 ng/mL (QC tested).