Learn More
ACROBiosystems ACROBiosystems Mouse Flt-3 / Flk-2 Protein, His Tag
Supplier:  ACROBiosystems FL3M52H61MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse Flt-3, His Tag (FL3-M52H6) is expressed from human 293 cells (HEK293). It contains AA Asn 28 - Ser 544 (Accession # NP_034359.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.9 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6630