Learn More
Enzo Life Sciences Flt-3 ligand (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX2018000100
Encompass_Preferred
Alternative name: FMS-related tyrosine kinase 3 ligand. Endotoxin: ≤1 EU/μg protein measured by kinetic LAL analysis. Purity: ≥95% (Reducing and Non-reducing SDS-PAGE). MW: ~17.6kDa. Formulation: Lyophilized from sterile filtered solution containing 10 mM sodium phosphate, 50 mM NaCl, pH 7.5. Source: Produced in E. coli. Non-glycosylated protein, containing 155 amino acids. Long Term Storage: -20°C.
Catalog No. 50-200-7991